missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PVR (CD155) Recombinant Mouse Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 345) of mouse PVR precursor (NP_081790.1) precursor fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$246.00 - $1,144.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4
Gene ID (Entrez) 52118
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50259M03H25 Life Technologies
LS50259M03H25
5 x 5μg Each for $246.00
50259M03H50 Life Technologies
LS50259M03H50
50μg Each for $374.00
50259M03HL Life Technologies
LS50259M03H250
5 x 50μg Each for $1,144.00
Specifications

Description

Poliovirus receptor (PVR/CD155), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse DNAM1 at 2μg/mL (100μL/well) can bind mouse PVR with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 50mM Tris, 10mM NaCl, pH 7.5, 100mM Glycine, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
52118
<1 EU/μg
3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4