Please login to your online account to display your discounted pricing

Sino Biological™ PVR (CD155) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 343) of human PVR (NP_006496.3) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10109H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10109H02H50

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Molecular Weight 61.8kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Viral Host Cell Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4
Common Name Poliovirus receptor
Gene Symbol PVR
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 5817

Poliovirus receptor (PVR/CD155), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human DNAM1 at 2μg/mL (100μL/well) can bind human PVR-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.