missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PVR (CD155) Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 343) of human PVR (NP_006496.3) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10109H02H250

 View more versions of this product

Catalog No. 10109H02HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; PVR (CD155) Recombinant Human Protein, hIgG1-Fc Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Poliovirus receptor (PVR/CD155), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human DNAM1 at 2μg/mL (100μL/well) can bind human PVR-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5817
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Host Cell Receptors
PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4
PVR
CD Proteins
IgG1 Fc Tag
Lyophilized
61.8kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Poliovirus receptor
Assay Standard
Mammalian