missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PVR (CD155) Recombinant Human Protein, DDDDK Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 343) of human PVR (NP_006496.3) fused to a DDDDK tag at the N-terminus

$120.00 - $1,172.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4
Gene ID (Entrez) 5817
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10109HCCH25 Life Technologies
LS10109HCCH25
5 x 5μg Each for $238.00
10109HCCH5 Life Technologies
LS10109HCCH5
5μg Each for $120.00
10109HCCHL Life Technologies
LS10109HCCH250
5 x 50μg Each for $1,172.00
10109HCCH50 Life Technologies
LS10109HCCH50
50μg Each for $381.00
Specifications

Description

Poliovirus receptor (PVR/CD155), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PVR (400ng/well) onto a mouse anti-human PVR antibody coated plate binds recombinant human CD226 with a linear range of 4.0-20μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
5817
<1 EU/μg
PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4