missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PTPRJ Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PTPRJ (Q12913) cytoplasmic domain (Arg 997-Ala 1337) fused to a polyhistidine tag at the N-terminus

$237.00 - $670.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
13165H07E25 Life Technologies
LS13165H07E25
5 x 5μg Each for $670.00
13165H07E5 Life Technologies
LS13165H07E5
5μg Each for $237.00
Specifications

Description

Protein Tyrosine Phosphatase, Receptor Type, J (PTPRJ), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1
PAGE
Lyophilized
5795