Please login to your online account to display your discounted pricing

Sino Biological™ PTPRJ Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human PTPRJ (Q12913) cytoplasmic domain (Arg 997-Ala 1337) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS13165H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13165H07E5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 41kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Truncated/Recombinant
Protein Subtype Tyrosine Phosphatases
Endotoxin Concentration <1 EU/μg
Gene Alias CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1
Common Name Protein tyrosine phosphatase, receptor type, J
Gene Symbol PTPRJ
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 5795

Protein Tyrosine Phosphatase, Receptor Type, J (PTPRJ), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.