Please login to your online account to display your discounted pricing

Sino Biological™ PTPN2 Recombinant Mouse Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$90.75 - $201.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias AI325124, Ptpt, TC-PTP
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50800M07B25

DFS Item
 
life technologies
LS50800M07B25
5 x 5μg Each for $201.00

50800M07B5

DFS Item
 
life technologies
LS50800M07B5
5μg Each for $90.75
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias AI325124, Ptpt, TC-PTP

Protein Tyrosine Phosphatase Nonreceptor Type 2 (PTPN2), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (988-998 phosphopeptide substrate, R&D Systems Cat. No. ES006); the specific activity is >15nM/min/μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, pH 8.0, 20% glycerol, 0.1mM TCEP
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.