Please login to your online account to display your discounted pricing

Sino Biological™ PTPN2 Recombinant Human Protein, N-His.GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$92.75 - $92.75

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10570H20B5

DFS Item
 
life technologies
LS10570H20B5
5μg Each for $92.75
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP

Protein Tyrosine Phosphatase Nonreceptor Type 2 (PTPN2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor 988-998 phosphopeptide substrate (R&D Systems Cat. No. ES006), the specific activity is >30nM/min./μg
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.