Please login to your online account to display your discounted pricing

Sino Biological™ PTPN1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human PTPN1 (NP_002818.1) (Glu 2 -Asn 321) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10304H07E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10304H07E50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 38kDa
Purity ≥95% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias PTP1B, PTPN1
Common Name Protein tyrosine phosphatase, nonreceptor type 3
Gene Symbol PTPN1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Protein tyrosine phosphatase, nonreceptor type 1 (PTPN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (aa 988-998) phosphopeptide substrate, the specific activity is >15nM/min./μg
  • Formulation: Lyophilized in 10mM HEPES, 500mM NaCl, 1mM DTT, 1mM EDTA, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.