missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PRMT5 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PRMT5 (NP_006128.1) (Ala 2-Leu 637) fused to a polyhistidine tag at the C-terminus and a Flag tag at the N-terminus

$141.00 - $542.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias PRMT5, HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11074H18H25 Life Technologies
LS11074H18H25
5 x 5μg Each for $357.00
11074H18H5 Life Technologies
LS11074H18H5
5μg Each for $141.00
11074H18H50 Life Technologies
LS11074H18H50
50μg Each for $542.00
Specifications

Description

Protein Arginine Methyltransferase 5 (PRMT5), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM EDTA, 0.5 PMSF, 3mM TCEP, 25% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
PRMT5, HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
PAGE
Frozen
10419