Please login to your online account to display your discounted pricing

Sino Biological™ PRMT5 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human PRMT5 (NP_006128.1) (Ala 2-Leu 637) fused to a polyhistidine tag at the C-terminus and a Flag tag at the N-terminus

Manufacturer: life technologies  LS11074H18H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11074H18H50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Format Frozen
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Gene ID (Entrez) 10419
Purity >85% by SDS-PAGE
Protein Family Methylation & Demethylation Proteins
Structural Form Full Length/Recombinant
Gene Alias PRMT5, HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
Gene Symbol PRMT5
Protein Tag N-Flag C-His
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 75kDa
Validated Application Assay Standard
Common Name Protein Arginine Methyltransferase 5
Purification Method PAGE
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Protein Arginine Methyltransferase 5 (PRMT5), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM EDTA, 0.5 PMSF, 3mM TCEP, 25% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.