missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PRMT5 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PRMT5 (NP_006128.1) (Ala 2-Leu 637) fused to a polyhistidine tag at the C-terminus and a Flag tag at the N-terminus

Manufacturer:  Life Technologies LS11074H18H25

 View more versions of this product

Catalog No. 11074H18H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; PRMT5 Recombinant Human Protein, His Tag; 5 x 5ug; Source: Human; Purity: greater than 85% by SDS-PAGE

Protein Arginine Methyltransferase 5 (PRMT5), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM EDTA, 0.5 PMSF, 3mM TCEP, 25% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10419
PAGE
5 x 5μg
Full Length/Recombinant
PRMT5, HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
PRMT5
Methylation & Demethylation Proteins
N-Flag C-His
Frozen
75kDa
>85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Protein Arginine Methyltransferase 5
Assay Standard
Mammalian