missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PRDX1 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse PRDX1 (P35700) (Met 1-Lys 199) fused to a polyhistidine tag at the C-terminus

$159.00 - $159.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias RP23-177C18.2, MSP23, NkefA, OSF-3, OSF3, PAG, Paga, PrdxI, PrxI, TDX2, TPxA, Tdpx2, prx1
Gene ID (Entrez) 18477
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50552M08E5 Life Technologies
LS50552M08E5
5μg Each for $159.00
Specifications

Description

Peroxiredoxin 1 (PRDX1), recombinant mouse protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: PBS, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
Frozen
18477
PAGE
RP23-177C18.2, MSP23, NkefA, OSF-3, OSF3, PAG, Paga, PrdxI, PrxI, TDX2, TPxA, Tdpx2, prx1