missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PPIA Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PPIA (P62937) (Met 1-Glu 165) fused to a polyhistidine tag at the C-terminus

$205.00 - $205.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias CYPA, CYPH, mgC117158, mgC12404, mgC23397
Gene ID (Entrez) 5478
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10436H08E25 View Documents Life Technologies
LS10436H08E25
5 x 5μg Each for $205.00
 
Description

Description

Peptidylprolyl Isomerase A (Cyclophin A) (PPIA), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
5478
PAGE
CYPA, CYPH, mgC117158, mgC12404, mgC23397

For Research Use Only. Not for use in diagnostic procedures.