Please login to your online account to display your discounted pricing

Sino Biological™ PPIA Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human PPIA (P62937) (Met 1-Glu 165) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10436H08E25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10436H08E25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 19.4kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Gene Alias CYPA, CYPH, mgC117158, mgC12404, mgC23397
Common Name Peptidylprolyl Isomerase A (Cyclophin A)
Gene Symbol PPIA
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene ID (Entrez) 5478

Peptidylprolyl Isomerase A (Cyclophin A) (PPIA), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.