missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ POSTN Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse POSTN (NP_056599.1) (Met 1-Gln 811) fused to a polyhistidine tag at the C-terminus

$216.00 - $354.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI747096, OSF-2, Osf2, PLF, PN, Periostin, peri
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50450M08H25 Life Technologies
LS50450M08H25
5 x 5μg Each for $216.00
50450M08H50 Life Technologies
LS50450M08H50
50μg Each for $354.00
Specifications

Description

Periostin, Osteoblast Specific Factor (POSTN), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
AI747096, OSF-2, Osf2, PLF, PN, Periostin, peri
PAGE
Lyophilized
50706