Please login to your online account to display your discounted pricing

Sino Biological™ PLAUR (UPAR) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human UPAR isoform 1 (Q03405-1) (Met 1-Arg 303) without the pro peptide, fused to the polyhistidine tag at the C-terminus

$92.75 - $806.00

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD87, U-PAR, UPAR, URKR
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10925H08H5

DFS Item
 
life technologies
LS10925H08H5
5μg Each for $92.75

10925H08H25

DFS Item
 
life technologies
LS10925H08H25
5 x 5μg Each for $202.00

10925H08H50

DFS Item
 
life technologies
LS10925H08H50
50μg Each for $259.00

10925H08HL

DFS Item
 
life technologies
LS10925H08H250
5 x 50μg Each for $806.00
Description & Specifications

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD87, U-PAR, UPAR, URKR

Plasminogen Activator, Urokinase Receptor (UPAR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human UPAR at 5μg/μL (100μL/well) can bind biotinylated human UPA with a linear ranger of 40-1000ng/μL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.