Please login to your online account to display your discounted pricing

Sino Biological™ PLAU Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PLAU (NP_002649.1) (Met 1-Leu 431) fused to the polyhistidine tag at the C-terminus

$175.00 - $2,090.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PLAU, ATF, UPA, URK, u-PA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10815H08H25

DFS Item
 
life technologies
LS10815H08H25
5 x 5μg Each for $512.00

10815H08H5

DFS Item
 
life technologies
LS10815H08H5
5μg Each for $175.00

10815H08H50

DFS Item
 
life technologies
LS10815H08H50
50μg Each for $752.00

10815H08HL

DFS Item
 
life technologies
LS10815H08H250
5 x 50μg Each for $2,090.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PLAU, ATF, UPA, URK, u-PA

Plasminogen Activator, Urokinase (PLAU), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PLAU at 5μg/mL (100μL/well) can bind mouse PLAUR with a linear range of 1.6-40ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.