Please login to your online account to display your discounted pricing

Sino Biological™ PLAU Recombinant Human Protein Activated by Trypsin, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PLAU (NP_002649.1) (Met 1-Leu 431) fused to the polyhistidine tag at the C-terminus

$167.00 - $167.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PLAU, ATF, UPA, URK, u-PA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10815H08HA5

DFS Item
 
life technologies
LS10815H08HA5
5μg Each for $167.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PLAU, ATF, UPA, URK, u-PA

Plasminogen Activator, Urokinase (PLAU) activated by trypsin, recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • The purified protein was activated by trypsin in vitro
  • Activity: Measured by its ability to cleave a peptide substrate, N-carbobenzyloxy-Gly-Gly-Arg-7-amido-4-methylcoumarin (Z-GGR-AMC), the specific activity is >2000pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.