missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PI3 (ELAFIN) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human PI3 (NP_002629.1) (Met 1-Gln 117) fused to a polyhistidine tag at the C-terminus
Manufacturer: Invitrogen LS12187H08H5
Description
Peptidase Inhibitor 3, Skin-Derived (PI3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Mammalian | |
Assay Standard | |
Lyophilized | |
11.3kDa | |
≥90% by SDS-PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Full Length/Recombinant | |
ESI, mgC13613, SKALP, WAP3, WFDC14, cementoin | |
PI3 |
Proteases, Peptidases & Inhibitors | |
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
5266 | |
PAGE | |
5μg | |
C-Terminal | |
<1 EU/μg | |
Peptidase inhibitor 3, skin-derived | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.