missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PDGFRB Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 531) of human PDGFRB (NP_002600.1) fused to a polyhistidine tag at the C-terminus

$189.00 - $189.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PDGFRB, CD140B, JTK12, PDGFR, PDGFR1
Gene ID (Entrez) 5159
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10514H08H25 View Documents Life Technologies
LS10514H08H25
5 x 5μg Each for $189.00
 
Description

Description

Platelet-derived growth factor receptor, beta polypeptide (PDGFRB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
5159
<1 EU/μg
PDGFRB, CD140B, JTK12, PDGFR, PDGFR1

For Research Use Only. Not for use in diagnostic procedures.