Please login to your online account to display your discounted pricing

Sino Biological™ PDGFRB Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 531) of human PDGFRB (NP_002600.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10514H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10514H08H50

  • / Each
 

Description & Specifications

Specifications

KinaseFamily PDGFR Kinase Family
Quantity 50μg
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 5159
Format Lyophilized
Gene Alias PDGFRB, CD140B, JTK12, PDGFR, PDGFR1
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥85% by SDS-PAGE
Protein Tag His Tag
Common Name Platelet-derived growth factor receptor, beta polypeptide
Gene Symbol PDGFRB
Protein Family Growth Factors & Receptors/CD Proteins
Expression System Mammalian
Structural Form Truncated/Recombinant
Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
Molecular Weight 57.6kDa
Validated Application Assay Standard

Platelet-derived growth factor receptor, beta polypeptide (PDGFRB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.