missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PDGFRA Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 524) of human PDGFRA (NP_006197.1) fused to a polyhistidine tag at the C-terminus

$355.00 - $355.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PDGFRA, CD140a, mgC74795, PDGFR2, Rhe-PDGFRA
Gene ID (Entrez) 5156
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10556H08H50 View Documents Invitrogen
LS10556H08H50
50μg Each for $355.00
 
Description

Description

Platelet-derived growth factor receptor, alpha polypeptide (PDGFRA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDGFRA-His at 1μg/well can bind human PDGFC-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
5156
<1 EU/μg
PDGFRA, CD140a, mgC74795, PDGFR2, Rhe-PDGFRA

For Research Use Only. Not for use in diagnostic procedures.