Please login to your online account to display your discounted pricing

Sino Biological™ PDGFRA Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 524) of human PDGFRA (NP_006197.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10556H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10556H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 57.7kDa
Purity ≥85% by SDS-PAGE
Format Lyophilized
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
Validated Application Assay Standard
Expression System Mammalian
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol PDGFRA
Endotoxin Concentration <1 EU/μg
Quantity 5μg
Gene ID (Entrez) 5156
Gene Alias PDGFRA, CD140a, mgC74795, PDGFR2, Rhe-PDGFRA
Protein Tag His Tag
Protein Family Growth Factors & Receptors/CD Proteins
Structural Form Truncated/Recombinant
Common Name Platelet-derived growth factor receptor, alpha
KinaseFamily PDGFR Kinase Family

Platelet-derived growth factor receptor, alpha polypeptide (PDGFRA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDGFRA-His at 1μg/well can bind human PDGFC-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.