Please login to your online account to display your discounted pricing

Sino Biological™ PDGFC Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

$135.00 - $1,740.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 1110064L01Rik, AI647969, PDGF-C
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50458M01H25

DFS Item
 
life technologies
LS50458M01H25
5 x 5μg Each for $349.00

50458M01H5

DFS Item
 
life technologies
LS50458M01H5
5μg Each for $135.00

50458M01HL

DFS Item
 
life technologies
LS50458M01H250
5 x 50μg Each for $1,740.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 1110064L01Rik, AI647969, PDGF-C

Platelet-derived growth factor C (PDGFC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: 1. Measured by its binding ability in a functional ELISA, immobilized mouse PDGFC (100μL/well of a 10μg/mL solution) can bind human PDGFRA; 2. Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.