Please login to your online account to display your discounted pricing

Sino Biological™ PDGFC Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human PDGFC (NP_057289.1) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

$109.00 - $1,434.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FALLOTEIN, SCDGF
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10273H01H25

DFS Item
 
life technologies
LS10273H01H25
5 x 5μg Each for $321.00

10273H01H5

DFS Item
 
life technologies
LS10273H01H5
5μg Each for $109.00

10273H01H50

DFS Item
 
life technologies
LS10273H01H50
50μg Each for $614.00

10273H01HL

DFS Item
 
life technologies
LS10273H01H250
5 x 50μg Each for $1,434.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FALLOTEIN, SCDGF

Platelet-derived growth factor C (PDGFC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.