Please login to your online account to display your discounted pricing

Sino Biological™ PDCD1LG2 Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met 1-Arg 219) fused to the Fc region of human IgG1 at the C-terminus

$812.00 - $812.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias B7-DC, Btdc, F730015O22Rik, mgC124039, mgC124040, PD-L2B7-DC, Btdc, F730015O22Rik, PD-L2
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50804M02HL

DFS Item
 
life technologies
LS50804M02H250
5 x 50μg Each for $812.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias B7-DC, Btdc, F730015O22Rik, mgC124039, mgC124040, PD-L2B7-DC, Btdc, F730015O22Rik, PD-L2

Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDCD1-His (Mfr. No. 10377-H08H) at 10μg/mL (100μL/well) can bind mouse PDCD1LG2-Fc; the EC50 of mouse PDCD1LG2-Fc is 0.95μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.