Please login to your online account to display your discounted pricing

Sino Biological™ PDCD1LG2 Recombinant Human Protein His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PDCD1LG2 (NP_079515.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10292H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10292H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 24kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias PDCD1LG2, B7DC, Btdc, PDL2, CD273, PD-L2, PDCD1L2, mgC142238, mgC142240, bA574F11.2
Common Name Programmed cell death 1 ligand 2
Gene Symbol PDCD1LG2
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human PDCD1LG2/PDL2 at 1μg/mL (100μL/well) can bind recombinant human PD1 with a linear range of 7.8-1000ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.