missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PDCD1LG2 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PDCD1LG2 (NP_079515.2) fused to the His-tagged Fc region of human IgG1 at the C-terminus

$225.00 - $225.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PDCD1LG2, B7DC, Btdc, PDL2, CD273, PD-L2, PDCD1L2, mgC142238, mgC142240, bA574F11.2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10292H03H25 View Documents Life Technologies
LS10292H03H25
5 x 5μg Each for $225.00
 
Description

Description

Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PD1 at 2μg/mL (100μg/well) can bind human PDCD1LG2/PDL2 with a linear range of 0.0064-0.16μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
PDCD1LG2, B7DC, Btdc, PDL2, CD273, PD-L2, PDCD1L2, mgC142238, mgC142240, bA574F11.2

For Research Use Only. Not for use in diagnostic procedures.