missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PCSK9 Recombinant Rat Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the rat PCSK9 (NP_954862.2) (Met 1-Gln 691) fused to a polyhistidine tag at the N-terminus

$247.00 - $247.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias NARC-1, Narc1, PC9, PCSK9
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
80005R08H5 View Documents Life Technologies
LS80005R08H5
5μg Each for $247.00
 
Description

Description

Proprotein convertase subtilisin/kexin type 9 (PCSK9), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind biotinylated human LDLR in a functional ELISA; 2. Measured by its ability to bind biotinylated mouse LDLR in a functional ELISA
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
NARC-1, Narc1, PC9, PCSK9

For Research Use Only. Not for use in diagnostic procedures.