missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PCSK9 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse PCSK9 (NP_705793.1) precursor (Met 1-Gln 694) fused to a C-terminal polyhistidine tag

$1,230.00 - $1,230.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias RP23-159J7.1, AI415265, AI747682, FH3, HCHOLA3, mgC47409, Narc1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50251M08H50 View Documents Life Technologies
LS50251M08H50
50μg Each for $1,230.00
 
Description

Description

Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse PCSK9 at 10μg/mL (100μL/well) can bind biotinylated recombinant human LDLR; the EC50 of biotinylated human LDLR is 0.12μg/mL
  • Formulation: 15mM Tris, 90mM NaCl, 50% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Frozen
RP23-159J7.1, AI415265, AI747682, FH3, HCHOLA3, mgC47409, Narc1

For Research Use Only. Not for use in diagnostic procedures.