Please login to your online account to display your discounted pricing

Sino Biological™ PCSK9 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PCSK9 (NP_777596.2) (Met 1-Gln 692) fused to a polyhistidine tag at the C-terminus

$239.00 - $4,115.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PCSK9, FH3, NARC1, LDLCQ1, NARC-1, HCHOLA3
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10594H08H25

DFS Item
 
life technologies
LS10594H08H25
5 x 5μg Each for $700.00

10594H08H5

DFS Item
 
life technologies
LS10594H08H5
5μg Each for $239.00

10594H08H50

DFS Item
 
life technologies
LS10594H08H50
50μg Each for $1,152.00

10594H08HL

DFS Item
 
life technologies
LS10594H08H250
5 x 50μg Each for $4,115.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PCSK9, FH3, NARC1, LDLCQ1, NARC-1, HCHOLA3

Proprotein convertase subtilisin/kexin type 9 (PCSK9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measure by its ability to bind with human LDLR in a functional ELISA; immobilized recombinant human PCSK9 at 10μg/mL (100μL/well) can bind biotinylated human LDLR, the EC50 of biotinylated human LDLR is 0.61μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.