missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ PCSK1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human PCSK1 (NP_000430.3) (Met 1-Arg 617) fused to a polyhistidine tag at the C-terminus

$682.00 - $682.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias PCSK1, BMIQ12, NEC1, PC1, PC3, SPC3
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10129H08H25 Life Technologies
LS10129H08H25
5 x 5μg Each for $682.00
Specifications

Description

Proprotein Convertase Subtilisin/Kexin Type 1 (PCSK1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
PCSK1, BMIQ12, NEC1, PC1, PC3, SPC3
PAGE
Lyophilized
5122