Please login to your online account to display your discounted pricing

Sino Biological™ PARP1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from the amino acids corresponding to the full length of human PARP1 (NP_001609.2) (Met 1-Trp 1014) fused to a polyhistidine tag at the C-terminus

$225.00 - $1,050.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11040H08B25

DFS Item
 
life technologies
LS11040H08B25
5 x 5μg Each for $666.00

11040H08B5

DFS Item
 
life technologies
LS11040H08B5
5μg Each for $225.00

11040H08B50

DFS Item
 
life technologies
LS11040H08B50
50μg Each for $1,050.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1

Poly (Adp-Ribose) Polymerase 1 (PARP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PARP1 at 10μg/mL (100μL/well) can bind to biotinylated human HSP70, the EC50 of biotinylated human HSP70 is 0.035μg/mL
  • Formulation: 50mM Tris, 100mM NaCl, pH 8, 2mM EDTA, 2mM PMSF, 0.5mM TCEP, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.