Please login to your online account to display your discounted pricing

Sino Biological™ OSMR Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human OSMR (NP_003990.1) extracellular domain (Met 1-Met 740) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11226H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11226H08H25

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Common Name Oncostatin M receptor
Gene ID (Entrez) 9180
Purity ≥90% by SDS-PAGE
Protein Tag His Tag
Molecular Weight 82.6kDa
Structural Form Truncated/Recombinant
Format Lyophilized
Purification Method PAGE
Gene Alias MGC150626, mgC150627, mgC75127, OSMRB
Protein Family Cytokines & Receptors
Termination C-Terminal
Validated Application Assay Standard
Gene Symbol OSMR
Expression System Mammalian
Endotoxin Concentration <1 EU/μg

Oncostatin M Receptor (OSMR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.