Please login to your online account to display your discounted pricing

Sino Biological™ OLR1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human OLR1 (P78380) extracellular domain (Ser 61-Gln 273) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10585H07H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10585H07HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 26.5kDa
Purification Method PAGE
Purity >85% by SDS-PAGE
Quantity 5 x 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias OLR1, CLEC8A, LOX1, LOXIN, SCARE1, SLOX1
Common Name Oxidized low density lipoprotein (lectin-like) receptor 1
Gene Symbol OLR1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 4973

Oxidized Low Density Lipoprotein (Lectin-like) Receptor 1 (OLR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.