Please login to your online account to display your discounted pricing

Sino Biological™ NRXN3 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human NRXN3 (NP_620426.2) extracellular domain (Met 1-Thr 357) fused to a polyhistidine tag at the C-terminus

$199.50 - $199.50

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias C14orf60, KIAA0743, mgC176711
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11843H08H25

DFS Item
 
life technologies
LS11843H08H25
5 x 5μg Each for $199.50
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias C14orf60, KIAA0743, mgC176711

Neurexin-3-Beta (NRXN3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; when 5 x 104 cells/well are added to NRXN3 coated plates (0.8μg/mL and 100μL/well), approximately 30-50% of the added cells will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.