missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NRP2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Tyr 855) of human NRP2 (NP_003863.2) fused to a C-terminal polyhistidine tag

$640.00 - $640.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias NRP2, mgC126574, NP2, NPN2, PRO2714, VEGF165R2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10695H08H50 View Documents Life Technologies
LS10695H08H50
50μg Each for $640.00
 
Description

Description

Neuropilin 2 (NRP2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human Neuropilin-2 at 2.5μg/mL (100μL/well) can bind recombinant human NRP2, the EC50 for this effect is typically 0.57nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
NRP2, mgC126574, NP2, NPN2, PRO2714, VEGF165R2

For Research Use Only. Not for use in diagnostic procedures.