Please login to your online account to display your discounted pricing

Sino Biological™ NRP1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human NRP1 isoform b (NP_001019799.1) (Met 1-Lys 644) fused to the polyhistidine tag at the C-terminus

$140.00 - $2,145.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-342D11.1, BDCA4, CD304, DKFZp686A03134, DKFZp781F1414, NP1, NRP1, VEGF165R
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10011H08H25

DFS Item
 
life technologies
LS10011H08H25
5 x 5μg Each for $378.00

10011H08H5

DFS Item
 
life technologies
LS10011H08H5
5μg Each for $140.00

10011H08HL

DFS Item
 
life technologies
LS10011H08H250
5 x 50μg Each for $2,145.00

10011H08H50

DFS Item
 
life technologies
LS10011H08H50
50μg Each for $600.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-342D11.1, BDCA4, CD304, DKFZp686A03134, DKFZp781F1414, NP1, NRP1, VEGF165R

Neutropilin-1 (NRP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by using the Octet RED System, the affinity constant (Kd) of NRP1-His bound to biotinylated human VEGF165 was 25nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.