missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ NRG4 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human NRG4 (Q8WWG1) extracellular domain (Pro 2-Phe 62) fused to additional two amino acids (Gly & Pro) at the N-terminus

$141.00 - $530.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HRG4
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12183HNCE5 Life Technologies
LS12183HNCE5
5μg Each for $141.00
12183HNCE50 Life Technologies
LS12183HNCE50
50μg Each for $530.00
Specifications

Description

Neuregulin 4 (NRG4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 50mM NaCl, pH 7.2, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
HRG4
PAGE
Lyophilized
145957