missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NRG4 Recombinant Human Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human NRG4 (Q8WWG1) extracellular domain (Pro 2-Phe 62) fused to additional two amino acids (Gly & Pro) at the N-terminus

Manufacturer:  Invitrogen LS12183HNCE50

 View more versions of this product

Catalog No. 12183HNCE50



Description

Description

Neuregulin 4 (NRG4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 50mM NaCl, pH 7.2, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Lyophilized
6.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Neuregulin 1
Growth Factors & Receptors/Cell Structure & Organelle Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
145957
PAGE
50μg
Truncated/Recombinant
HRG4
NRG4
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.