Please login to your online account to display your discounted pricing

Sino Biological™ NRG1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human NRG1 isoform alpha (Q02297-1) (Ser 20-Lys 241) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS13499H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13499H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 25.5kDa
Gene Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF
Protein Tag His Tag
Validated Application Assay Standard
Expression System Mammalian
Quantity 5μg
Purity ≥95% by SDS-PAGE
Gene Symbol NRG1
Format Lyophilized
Common Name Neuregulin 1
Endotoxin Concentration <1 EU/μg
Protein Family Growth Factors & Receptors
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Growth Factors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Neuregulin 1 (NRG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.