Please login to your online account to display your discounted pricing

Sino Biological™ NRG1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the EGF-like domain (Thr 176-Lys 246) of human NRG1 (Q02297-6) fused to the Fc region of human IgG1 at the N-terminus

$91.00 - $210.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11609H01H25

DFS Item
 
life technologies
LS11609H01H25
5 x 5μg Each for $210.00

11609H01H5

DFS Item
 
life technologies
LS11609H01H5
5μg Each for $91.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF

Neuregulin 1 (NRG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized Rhesus ErbB3 at 2μg/mL (100μL/well) can bind human NRG1 (isoform Beta1), the EC50 of human NRG1 (isoform Beta1) is typically 0.58μg/mL; 2. Immobilized human ErbB3 at 2μg/mL (100μL/well) can bind human NRG1 (isoform Beta1), the EC50 of human NRG1 (isoform Beta1) is typically 0.43μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.