missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NOV Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of human NOV (NP_002505.1) (Met 1-Met 357) fused to a polyhistidine tag at the C-terminus

$94.75 - $94.75

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias NOV, CCN3, IGFBP9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10264H08B5 View Documents Life Technologies
LS10264H08B5
5μg Each for $94.75
 
Description

Description

Nephroblastoma Overexpressed (NOV), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Balb/3T3 mouse embryonic fibroblast cells; when cells are added to NOV coated plates (10μg/mL, 100μL/well), 50%-60% will adhere specifically after 60 minutes at 37°C
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 10mM Imidazole, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Frozen
NOV, CCN3, IGFBP9

For Research Use Only. Not for use in diagnostic procedures.