missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ NOV Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of human NOV (NP_002505.1) (Met 1-Met 357) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10264H08B5

 View more versions of this product

Catalog No. 10264H08B5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; NOV Recombinant Human Protein, His Tag; 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Nephroblastoma Overexpressed (NOV), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Balb/3T3 mouse embryonic fibroblast cells; when cells are added to NOV coated plates (10μg/mL, 100μL/well), 50%-60% will adhere specifically after 60 minutes at 37°C
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 10mM Imidazole, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37kDa
5μg
Other Proteins
NOV, CCN3, IGFBP9
NOV
Insect
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Nephroblastoma overexpressed
Assay Standard
His Tag