missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ NGF Recombinant Mouse Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse NGF (NP_001106168.1), residues 122 to 241, with an initial Met at the N-terminus

$65.75 - $770.00

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Ngf, Ngfb
Gene ID (Entrez) 18049
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50385MNAC25 Life Technologies
LS50385MNAC25
5 x 5μg Each for $180.00
50385MNAC5 Life Technologies
LS50385MNAC5
5μg Each for $65.75
50385MNAC50 Life Technologies
LS50385MNAC50
50μg Each for $222.00
50385MNACL Life Technologies
LS50385MNAC250
5 x 50μg Each for $770.00
Specifications

Description

Nerve growth factor (NGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells; the ED50 for this effect is 0.2-2.0ng/mL
  • Formulation: Lyophilized in 50mM Na acetate buffer, pH 5.5, 150mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
18049
<1 EU/μg
Ngf, Ngfb