missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neuraminidase (NA) Recombinant Influenza A Virus Protein Subtype H3N2 (A/Babol/36/2005), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the Influenza A H3N2 virus (A/Babol/36/2005) Neuraminidase (ACN50232.1) (His 36-Pro 459) fused to a polyhistidine tag at the N-terminus

$146.00 - $146.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias NA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
40017V07H5 View Documents Life Technologies
LS40017V07H5
5μg Each for $146.00
 
Description

Description

Neuraminidase (NA), recombinant Influenza A H3N2 ( A/Babol/36/2005 ) NA/Neuraminidase protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
NA
PAGE
Lyophilized
Unknown

For Research Use Only. Not for use in diagnostic procedures.