Please login to your online account to display your discounted pricing

Sino Biological™ MUC1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human MUC1 isoform 2 (NP_001018016.1) extracellular domain (Met 1-Gly 167) fused to the Fc region of human IgG1 at the C-terminus

$105.00 - $344.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-263K19.2, CD227, EMA, H23AG, KL-6, MAM6, PEM, PEMT, PUM
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

12123H02H25

DFS Item
 
life technologies
LS12123H02H25
5 x 5μg Each for $225.00

12123H02H5

DFS Item
 
life technologies
LS12123H02H5
5μg Each for $105.00

12123H02H50

DFS Item
 
life technologies
LS12123H02H50
50μg Each for $344.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-263K19.2, CD227, EMA, H23AG, KL-6, MAM6, PEM, PEMT, PUM

Mucin 1 Cell Surface Associated (MUC1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated mouse ERBB2 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.