Please login to your online account to display your discounted pricing

Sino Biological™ mMP9 Recombinant Rat Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the ratmMP9 (EDL96479.1) (Met 1-Pro 708) fused to a polyhistidine tag at the C-terminus

$256.00 - $256.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias Mmp9
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

80049R08H5

DFS Item
 
life technologies
LS80049R08H5
5μg Each for $256.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias Mmp9

Matrix Metallopeptidase 9 (MMP9), recombinant rat protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (R&D Systems Cat. No. ES010); the specific activity is >1000pmol/min./μg
  • Formulation: 50mM MES, 100mM NaCl, 1mM CaCl2, 0.2μg/μL BSA, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.