missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

mMP9 Recombinant Mouse Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mousemMP9 (P41245-1) (Met 1-Pro 730)

$279.00 - $279.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-61O3.10-001, AW743869, B/MMP9, Clg4b, mMP-9, pro-MMP-9
View More Specs

Includes: Five vials of protein in suspension buffer

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50560MNAH5 View Documents Life Technologies
LS50560MNAH5
5μg Each for $279.00
 
Description

Description

Matrix Metallopeptidase 9 (MMP9), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP23-61O3.10-001, AW743869, B/MMP9, Clg4b, mMP-9, pro-MMP-9
PAGE
Lyophilized
17395

For Research Use Only. Not for use in diagnostic procedures.